Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248396.1 | NP_001235325.1 | 3 - 4 | 427 | 104 | |
GenBank mRNA | AK246074.1 | - | 3 - 4 | 468 | 104 | |
BT097673.1 | - | 3 - 4 | 465 | 104 | ||
EF521872.1 | - | 3 - 4 | 427 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |