Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK245832.1 | - | 3 - 4 | 1183 | 78 | |
EU019566.1 | - | 1 - 2 | 491 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |