Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244489.1 | - | 3 - 4 | 1157 | 104 | |
EU019576.1 | - | 1 - 2 | 372 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |