Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250504.1 | NP_001237433.1 | 6 - 7 | 1212 | 88 | |
GenBank mRNA | EU033956.1 | - | 6 - 7 | 1212 | 88 | |
KF359958.1 | - | 6 - 7 | 1212 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |