Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALAAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250624.1 | NP_001237553.1 | 3 - 4 | 268 | 101 | |
GenBank mRNA | EU165371.1 | - | 3 - 4 | 268 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |