Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250726.1 | NP_001237655.1 | 2 - 3 | 927 | 100 | |
GenBank mRNA | EU375355.1 | - | 2 - 3 | 927 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |