Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251781.2 | NP_001238710.1 | 6 - 7 | 1160 | 75 | |
GenBank mRNA | AB359233.1 | - | 6 - 7 | 1211 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |