Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249075.1 | NP_001236004.1 | 5 - 6 | 1084 | 88 | |
GenBank mRNA | FJ795040.1 | - | 5 - 6 | 1084 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Glycine max Probe spans exons |