Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248759.1 | NP_001235688.1 | 1 | 752 | 87 | |
GenBank mRNA | DQ822888.1 | - | 1 | 752 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |