Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012409.hg.1 |
TC04000912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012409.hg.1 |
ENST00000296795.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012409.hg.1 |
ENST00000542020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012409.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012408.hg.1 |
TC04000912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012408.hg.1 |
ENST00000296795.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012408.hg.1 |
ENST00000542020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012408.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000508051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000508051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000508051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000504367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000508051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000508051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000508051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000512264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009543.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165342.hg.1 |
TC0400009543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
ENST00000296795.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
ENST00000513189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
NM_003265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165344.hg.1 |
TC0400009543.hg.1 |
Clariom D TC |