Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000457564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000457564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000457564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021827.hg.1 |
TC06001807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021827.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021827.hg.1 |
ENST00000370963.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021827.hg.1 |
ENST00000486559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021827.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183028.hg.1 |
TC0600012059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000370963.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000457564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000457564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000457564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000477599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
ENST00000486559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012059.hg.1 |
NM_001512.3 |
Refseq |