Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020337.hg.1 |
TC0X001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020337.hg.1 |
ENST00000361930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020337.hg.1 |
ENST00000369926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020337.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020337.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020338.hg.1 |
TC0X001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020338.hg.1 |
ENST00000361930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020338.hg.1 |
ENST00000369926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020338.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020338.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011191.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176329.hg.1 |
TC0X00011191.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000610938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000617213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000617225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
ENST00000619903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
NM_001205201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
NM_001586.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176330.hg.1 |
TC0X00011191.hg.1 |
Clariom D TC |