Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017298.hg.1 |
TC04001269.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017298.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017298.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010944.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
ENST00000353151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
NM_001302770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
NM_001891.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174446.hg.1 |
TC0400010944.hg.1 |
Clariom D TC |