Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003102.2 | NP_003093.2 | 1 - 2 | 193 | 99 | |
GenBank mRNA | AK313935.1 | - | 1 - 2 | 174 | 99 | |
BC014418.1 | - | 1 - 2 | 58 | 4644224 | 99 | |
BI757365.1 | - | 1 - 2 | 192 | 99 | ||
DA668533.1 | - | 1 - 2 | 178 | 99 | ||
J02947.1 | - | 1 - 2 | 57 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Human Hypoxia Panel Human Antioxident Mechanisms 1700 Inventoried Human_Hypoxia Human_Antioxident_Mechanisms Probe spans exons |