Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024851.hg.1 |
ENST00000310473.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024851.hg.1 |
ENST00000536983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024851.hg.1 |
ENST00000542615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024851.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024851.hg.1 |
TC11002074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024850.hg.1 |
ENST00000310473.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024850.hg.1 |
ENST00000536983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024850.hg.1 |
ENST00000542615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024850.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024850.hg.1 |
TC11002074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024848.hg.1 |
ENST00000310473.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024848.hg.1 |
ENST00000536983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024848.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024848.hg.1 |
TC11002074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000545212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000545212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000545212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000545562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000545562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
ENST00000545562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189482.hg.1 |
TC1100011602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189489.hg.1 |
TC1100011602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189483.hg.1 |
TC1100011602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189490.hg.1 |
TC1100011602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000310473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000536983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000539330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000539764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000541027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000542615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000543714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000544615.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000545212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000545212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000545212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000545562.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000545562.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
ENST00000545562.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011602.hg.1 |
NM_003355.2 |
Refseq |