Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015359.hg.1 |
TC0X001128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015359.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015359.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015359.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015359.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015360.hg.1 |
TC0X001128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015360.hg.1 |
ENST00000373687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015360.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015360.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015360.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015360.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010005.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015363.hg.1 |
TC0X001128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015363.hg.1 |
ENST00000373687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015363.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015363.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015363.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015363.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168304.hg.1 |
TC0X00010005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010005.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
ENST00000373691.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
ENST00000373693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
NM_001142797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
NM_001504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168300.hg.1 |
TC0X00010005.hg.1 |
Clariom D TC |