Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020061.hg.1 |
TC09001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020061.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020061.hg.1 |
ENST00000373425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020061.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020064.hg.1 |
TC09001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020064.hg.1 |
ENST00000373425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020064.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020064.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000470194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000470194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178803.hg.1 |
TC0900011565.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000470194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000470194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011565.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
ENST00000470194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
ENST00000373425.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
ENST00000470194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
ENST00000491991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
NM_012098.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178806.hg.1 |
TC0900011565.hg.1 |
Clariom D TC |