Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011892.hg.1 |
TC08000906.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011892.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011892.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164548.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164548.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164548.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164548.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164548.hg.1 |
TC0800009429.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800164547.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164547.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164547.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164547.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164547.hg.1 |
TC0800009429.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
ENST00000297439.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009429.hg.1 |
NM_005218.3 |
Refseq |