Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000917.hg.1 |
ENST00000378961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000917.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000917.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000917.hg.1 |
TC20000062.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000914.hg.1 |
ENST00000378961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000914.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000914.hg.1 |
TC20000062.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000912.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000912.hg.1 |
TC20000062.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000913.hg.1 |
ENST00000378961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000913.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000913.hg.1 |
TC20000062.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000145443.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145443.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145443.hg.1 |
TC2000006627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145443.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145443.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006627.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
TC2000006627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145445.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145442.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145442.hg.1 |
TC2000006627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145442.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
TC2000006627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145444.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
NM_001819.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
TC2000006627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
ENST00000378961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
ENST00000455042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
ENST00000488832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145448.hg.1 |
NM_001819.2 |
Refseq |