Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000813.hg.1 |
TC08000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000813.hg.1 |
ENST00000259089.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000813.hg.1 |
ENST00000427279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000813.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000813.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000813.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000816.hg.1 |
TC08000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000816.hg.1 |
ENST00000259089.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000816.hg.1 |
ENST00000427279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000816.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000812.hg.1 |
TC08000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000812.hg.1 |
ENST00000259089.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000812.hg.1 |
ENST00000427279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000812.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000812.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000812.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000810.hg.1 |
TC08000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000810.hg.1 |
ENST00000259089.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000810.hg.1 |
ENST00000427279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000810.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000810.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000809.hg.1 |
TC08000087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000809.hg.1 |
ENST00000259089.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000809.hg.1 |
ENST00000427279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000809.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145691.hg.1 |
TC0800006746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145690.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145690.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145690.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145690.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145690.hg.1 |
TC0800006746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145694.hg.1 |
TC0800006746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145697.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145697.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145697.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145697.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145697.hg.1 |
TC0800006746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145693.hg.1 |
TC0800006746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000259089.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000525389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000526778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000529894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
ENST00000533828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006746.hg.1 |
NM_001715.2 |
Refseq |