Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014142.hg.1 |
ENST00000217185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014142.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014142.hg.1 |
TC20001033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014143.hg.1 |
ENST00000217185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014143.hg.1 |
ENST00000542869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014143.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014143.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014143.hg.1 |
TC20001033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
TC2000009815.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168527.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168526.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168526.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168526.hg.1 |
TC2000009815.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168526.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168526.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
TC2000009815.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168528.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000217185.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
ENST00000542869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_001256358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009815.hg.1 |
NM_005975.3 |
Refseq |