Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018977.hg.1 |
ENST00000301046.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018977.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018977.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018977.hg.1 |
TC12001451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018978.hg.1 |
ENST00000301046.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018978.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018978.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018978.hg.1 |
TC12001451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018979.hg.1 |
ENST00000301046.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018979.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018979.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018979.hg.1 |
TC12001451.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010585.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180071.hg.1 |
TC1200010585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180073.hg.1 |
TC1200010585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
ENST00000301046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
ENST00000549817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
NM_002289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180072.hg.1 |
TC1200010585.hg.1 |
Clariom D TC |