Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004578.3 | NP_004569.2 | 3 - 4 | 504 | 64 | |
NR_073545.1 | 3 - 4 | 504 | 64 | |||
GenBank mRNA | AF498934.1 | - | 3 - 4 | 226 | 64 | |
AK223009.1 | - | 3 - 4 | 269 | 64 | ||
AK313807.1 | - | 3 - 4 | 477 | 64 | ||
AY585832.1 | - | 3 - 4 | 433 | 64 | ||
BC002438.1 | - | 3 - 4 | 297 | 3346455 | 64 | |
BC004309.1 | - | 3 - 4 | 434 | 3627110 | 64 | |
BI828974.1 | - | 3 - 4 | 388 | 64 | ||
KC407675.1 | - | 3 - 4 | 226 | 64 | ||
M28211.1 | - | 3 - 4 | 281 | 64 | ||
X13902.1 | - | 3 - 4 | 337 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens HUMAN INSULIN RECEPTOR PATHWAY Inventoried Human_Insulin_Receptor_Pathway Probe spans exons |