Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053609.hg.1 |
TC01003421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053609.hg.1 |
ENST00000367996.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053609.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053609.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053608.hg.1 |
TC01003421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053608.hg.1 |
ENST00000367996.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053608.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223080.hg.1 |
TC0100016163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
ENST00000478394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016163.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223079.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223079.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223079.hg.1 |
ENST00000367996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223079.hg.1 |
NM_005099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223079.hg.1 |
TC0100016163.hg.1 |
Clariom D TC |