Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
ENST00000514815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000506544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
ENST00000514815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011538.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000502431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000512641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179211.hg.1 |
TC0400011538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000265174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000504987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000511304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
ENST00000514489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
NM_005443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179218.hg.1 |
TC0400011538.hg.1 |
Clariom D TC |