Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019619.hg.1 |
TC06001660.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019619.hg.1 |
ENST00000361288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019619.hg.1 |
ENST00000420584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019619.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019620.hg.1 |
TC06001660.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019620.hg.1 |
ENST00000361288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019620.hg.1 |
ENST00000420584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019620.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179560.hg.1 |
TC0600011600.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179562.hg.1 |
TC0600011600.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179559.hg.1 |
TC0600011600.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011600.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
ENST00000361288.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
ENST00000420584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
NM_001270488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
NM_005643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179558.hg.1 |
TC0600011600.hg.1 |
Clariom D TC |