Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011198.hg.1 |
ENST00000455429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010262.hg.1 |
ENST00000426049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011197.hg.1 |
ENST00000418167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011196.hg.1 |
ENST00000455013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011804.hg.1 |
ENST00000525775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011804.hg.1 |
ENST00000525775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010262.hg.1 |
ENST00000426049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011198.hg.1 |
ENST00000455429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011197.hg.1 |
ENST00000418167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011804.hg.1 |
ENST00000525775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011804.hg.1 |
ENST00000525775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011196.hg.1 |
ENST00000455013.2 |
Ensembl |