Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_006674_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118112 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118114 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118115 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118113 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8118111 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
206082_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11720923_PM_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11720923_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11720922_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11720922_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003473.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003473.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003476.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003476.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000890.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000890.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000682.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000682.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000685.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000685.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000889.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000889.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000681.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000681.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000687.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000687.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003472.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003472.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003478.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003478.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000686.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000686.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003475.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003475.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000893.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000893.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000698.hg.1 |
TC6_mcf_hap5000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000698.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003477.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003477.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000891.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000891.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000895.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000895.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000684.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000684.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000892.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000892.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003474.hg.1 |
TC06004072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003474.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000894.hg.1 |
TC6_cox_hap2000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000894.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000683.hg.1 |
TC6_ssto_hap7000048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000683.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198125.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198120.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198122.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014102.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000467369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198121.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000460889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198124.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198126.hg.1 |
ENST00000541196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198126.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198126.hg.1 |
ENST00000541196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198126.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198126.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
ENST00000541196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
NR_040662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198123.hg.1 |
TC0600014102.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
206082_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
206082_at |
- |
- |
Human Genome U133A 2.0 Array |
206082_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
206082_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_006674_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw526189 |
- |
- |
Human Genome U95 Set |
33320_PM_at |
- |
- |
Human Genome U95 Set |
33320_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206082_at7 |
- |
- |
Human Genome U133 Set |
206082_PM_at |
- |
- |
Human Genome U133 Set |
206082_at |
- |
- |