Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319720.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000319756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000391804.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000453757.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000593681.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000593681.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000593681.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000593681.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000593681.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000593681.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000594827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000598799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000600362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000601671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000601671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000601671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000601671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
ENST00000601671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001136032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_001167605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_006853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011234.hg.1 |
NM_144947.2 |
Refseq |