Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000601154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
ENST00000601154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009433.hg.1 |
ENST00000221922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009433.hg.1 |
ENST00000504556.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009433.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009433.hg.1 |
TC19000674.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
TC1900008415.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164659.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000221922.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000595659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000596938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000599398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000600117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000601154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
ENST00000601154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008415.hg.1 |
NM_015603.2 |
Refseq |