Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
ENST00000330442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
ENST00000342651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
ENST00000404360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017637.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
ENST00000330442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
ENST00000342651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
ENST00000404360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017639.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000491565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000491565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
ENST00000491565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
ENST00000330442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
ENST00000342651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
ENST00000404360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017635.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
ENST00000330442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
ENST00000342651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
ENST00000404360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017643.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006655.hg.1 |
ENST00000577401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
ENST00000342651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
ENST00000404360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017634.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
ENST00000330442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
ENST00000342651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
ENST00000404360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017641.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017640.hg.1 |
TC07001134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017640.hg.1 |
ENST00000330442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017640.hg.1 |
ENST00000399476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017640.hg.1 |
ENST00000405858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07017640.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172680.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172680.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172680.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172680.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172680.hg.1 |
TC0700010216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172681.hg.1 |
TC0700010216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006655.hg.1 |
ENST00000577401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000394917.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000491565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000491565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
ENST00000491565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010216.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172679.hg.1 |
TC0700010216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
ENST00000342651.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
ENST00000404360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
ENST00000405858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
NM_006956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
NM_016265.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172683.hg.1 |
TC0700010216.hg.1 |
Clariom D TC |