Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022365.hg.1 |
TC05001644.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022365.hg.1 |
ENST00000399004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022365.hg.1 |
ENST00000508629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022365.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022370.hg.1 |
TC05001644.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022370.hg.1 |
ENST00000399004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022370.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000511400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000511400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022374.hg.1 |
TC05001644.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022374.hg.1 |
ENST00000399004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022374.hg.1 |
ENST00000508629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022374.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022372.hg.1 |
TC05001644.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022372.hg.1 |
ENST00000399004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022372.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022373.hg.1 |
TC05001644.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022373.hg.1 |
ENST00000399004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022373.hg.1 |
ENST00000508629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022373.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000507478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000511400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000511400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011602.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180371.hg.1 |
TC0500011602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000399004.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000508629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000512248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000513603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
ENST00000513747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
NM_017676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180365.hg.1 |
TC0500011602.hg.1 |
Clariom D TC |