Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020679.hg.1 |
TC08001575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020679.hg.1 |
ENST00000259512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020679.hg.1 |
ENST00000419562.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020679.hg.1 |
ENST00000519018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020679.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020679.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179173.hg.1 |
TC0800011669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000259512.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000405944.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000419562.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000519018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000523036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
ENST00000524119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_001134671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011669.hg.1 |
NM_024295.5 |
Refseq |