Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013923.hg.1 |
TC07000895.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013923.hg.1 |
ENST00000247883.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013923.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013921.hg.1 |
TC07000895.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013921.hg.1 |
ENST00000247883.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013921.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166728.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166728.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166728.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166728.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166728.hg.1 |
TC0700009400.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009400.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166726.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166726.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166726.hg.1 |
ENST00000247883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166726.hg.1 |
NM_018980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166726.hg.1 |
TC0700009400.hg.1 |
Clariom D TC |