Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000474418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000474418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033715.hg.1 |
TC03003417.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033715.hg.1 |
ENST00000336685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033715.hg.1 |
ENST00000497985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033715.hg.1 |
ENST00000535500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033715.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033715.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000474418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000474418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000489015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000610787.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014061.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
ENST00000336685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
ENST00000497985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
ENST00000613069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
NM_001199978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
NM_001199979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
NM_020359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209174.hg.1 |
TC0300014061.hg.1 |
Clariom D TC |