Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000365128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000528833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000528833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000528833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000533680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000630083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
ENST00000630083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006810.hg.1 |
NR_002962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002311.hg.1 |
ENST00000379719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002311.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002311.hg.1 |
TC11000170.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000365128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000527431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000528833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000528833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000528833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000530037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000533680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000630083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
ENST00000630083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006810.hg.1 |
NR_002962.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
ENST00000379719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
ENST00000531235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
NM_006391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147747.hg.1 |
TC1100006810.hg.1 |
Clariom D TC |