Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010952.hg.1 |
ENST00000305628.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010952.hg.1 |
ENST00000317643.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010952.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010952.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010952.hg.1 |
TC19000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010951.hg.1 |
ENST00000305628.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010951.hg.1 |
ENST00000317643.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010951.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010951.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010951.hg.1 |
TC19000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010948.hg.1 |
ENST00000305628.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010948.hg.1 |
ENST00000317643.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010948.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010948.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010948.hg.1 |
TC19000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010949.hg.1 |
ENST00000305628.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010949.hg.1 |
ENST00000317643.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010949.hg.1 |
ENST00000536156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010949.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010949.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010949.hg.1 |
TC19000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010950.hg.1 |
ENST00000317643.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010950.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010950.hg.1 |
TC19000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010953.hg.1 |
ENST00000305628.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010953.hg.1 |
ENST00000317643.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010953.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010953.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010953.hg.1 |
TC19000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000536156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000536156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000536156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000599250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000599250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000599250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000600577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000600577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
TC1900008625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167394.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167389.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167389.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167389.hg.1 |
TC1900008625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167389.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167389.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000536156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000536156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000536156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000599250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000599250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000599250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000600577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000600577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_001277201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008625.hg.1 |
NR_102350.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
TC1900008625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
ENST00000601682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167390.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008628.hg.1 |
ENST00000595243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
TC1900008625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167393.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
NM_016543.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
TC1900008625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
ENST00000305628.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
ENST00000317643.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
NM_014385.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167395.hg.1 |
NM_016543.3 |
Refseq |