Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014324.hg.1 |
TC03000798.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014324.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014324.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000349243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000404754.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000418473.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000461609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000474935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000475347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
ENST00000497524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_000685.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_004835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_009585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_031850.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009119.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165342.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165342.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165342.hg.1 |
ENST00000402260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165342.hg.1 |
NM_032049.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165342.hg.1 |
TC0300009119.hg.1 |
Clariom D TC |