Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022452.hg.1 |
TC03001302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022452.hg.1 |
ENST00000273153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022452.hg.1 |
ENST00000318290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022452.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022452.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022454.hg.1 |
TC03001302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022454.hg.1 |
ENST00000273153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022454.hg.1 |
ENST00000318290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022454.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022455.hg.1 |
TC03001302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022455.hg.1 |
ENST00000318290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022455.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022451.hg.1 |
TC03001302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022451.hg.1 |
ENST00000273153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022451.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022451.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022453.hg.1 |
TC03001302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022453.hg.1 |
ENST00000273153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022453.hg.1 |
ENST00000318290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022453.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178531.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178531.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178531.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178531.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178531.hg.1 |
TC0300010770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178528.hg.1 |
TC0300010770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178530.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178530.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178530.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178530.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178530.hg.1 |
TC0300010770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178529.hg.1 |
TC0300010770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000273153.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
ENST00000514182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010770.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178532.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178532.hg.1 |
NM_033027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178532.hg.1 |
TC0300010770.hg.1 |
Clariom D TC |