Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005372.1 | NP_005363.1 | 1 | 390 | 96 | |
GenBank mRNA | BC069569.1 | - | 1 | 390 | 7262233 | 96 |
BC069590.1 | - | 1 | 390 | 96 | ||
BC106737.1 | - | 1 | 390 | 40016104 | 96 | |
BC106738.1 | - | 1 | 390 | 40016106 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |