Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
ENST00000241124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
ENST00000400065.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
ENST00000400066.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005199.hg.1 |
TC13000457.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155670.hg.1 |
TC1300008250.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000241124.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000356192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
ENST00000400066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110220.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_001110221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008250.hg.1 |
NM_006783.4 |
Refseq |