Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008992.hg.1 |
ENST00000225298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008992.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008992.hg.1 |
TC17000681.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161816.hg.1 |
TC1700008283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000225298.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000508506.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000579261.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000582832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
ENST00000583725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008283.hg.1 |
NM_016001.2 |
Refseq |