Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002072.4 | NP_002063.2 | 7 | 4426 | 70 | |
XM_017014628.1 | XP_016870117.1 | 7 | 4155 | 70 | ||
GenBank mRNA | AJ420425.1 | - | 1 | 117 | 70 | |
AK055619.1 | - | 1 | 252 | 70 | ||
BC047048.1 | - | 1 | 800 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |