Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
TC06002270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
ENST00000367066.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028488.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028487.hg.1 |
TC06002270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028487.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028487.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194715.hg.1 |
TC0600013743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600194713.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194713.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194713.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194713.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194713.hg.1 |
TC0600013743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000326965.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000338313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
ENST00000367066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_001278733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_054114.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_138810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013743.hg.1 |
NM_152133.2 |
Refseq |