Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021626.hg.1 |
ENST00000244934.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021626.hg.1 |
ENST00000378024.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021626.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021626.hg.1 |
TC11001867.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000525875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000525875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000525875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183855.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183855.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183855.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183855.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183855.hg.1 |
TC1100011083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100183856.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183856.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183856.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183856.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183856.hg.1 |
TC1100011083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000257247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000378024.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000525875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000525875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000525875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000528508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000530285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000531324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
ENST00000533365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_001620.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011083.hg.1 |
NM_024060.3 |
Refseq |