Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
ENST00000253001.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
ENST00000279068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
ENST00000370906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
ENST00000370915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
ENST00000444156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006720.hg.1 |
TC20000482.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007991.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
NM_144703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
TC2000007991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000279068.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000279069.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000361670.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000370915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
ENST00000400318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155543.hg.1 |
NM_144703.2 |
Refseq |