Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042617.hg.1 |
TC01002717.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042617.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042617.hg.1 |
ENST00000492633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042617.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207585.hg.1 |
TC0100014365.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000371204.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000466095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000468257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000469406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
ENST00000492633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014365.hg.1 |
NM_000775.3 |
Refseq |