Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023907.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023907.hg.1 |
ENST00000353903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023907.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023907.hg.1 |
TC11001988.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
ENST00000294288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
ENST00000353903.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
ENST00000545205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
NM_016366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187476.hg.1 |
TC1100011339.hg.1 |
Clariom D TC |