Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK000398.1 | - | 1 - 2 | 239 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons |