Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
TC04001300.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
ENST00000357854.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
ENST00000359707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
ENST00000395719.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
NM_012297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
NM_203504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017703.hg.1 |
NM_203505.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000357854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000359707.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000395719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000507133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
NM_012297.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
NM_203504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
NM_203505.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000357854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000359707.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000395719.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
ENST00000507133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
NM_012297.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
NM_203504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
NM_203505.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400190245.hg.1 |
TC0400012931.hg.1 |
Clariom D TC |